This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 2010107G23RIK SIRNA Assays ›
Gene Symbol:
2010107G23Rik
Gene Name:
RIKEN cDNA 2010107G23 gene
Gene Aliases:
AA545165
Chromosome Location:
Chr.10: 62107656 - 62111074 on Build GRCm38
UniGene ID:
Mm.294126
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 81578

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2010107G23Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027251.3 NP_081527.1 3, 4 271
GenBank mRNA AK008338.1 - 2, 3 191
AK035688.1 - 3, 4 272
BC024943.1 - 3, 4 586 3989515

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001127452, XM_006239781, BC158618.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0