This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAAP1 SIRNA Assays ›
Gene Symbol:
Caap1
Gene Name:
caspase activity and apoptosis inhibitor 1
Gene Aliases:
5830433M19Rik, AI536248, CAAP
Chromosome Location:
Chr.4: 94460277 - 94557076 on Build GRCm38
UniGene ID:
Mm.35170
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 80503

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Caap1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026368.2 NP_080644.2 2 413
GenBank mRNA AK013385.1 - 2 383
AK032696.1 - 2 383
AK046647.1 - 2 363
AK076487.1 - 2 368
AK140382.1 - 2 414
AK150311.1 - 3 323
AK160297.1 - 2 414
AK160572.1 - 2 414
AK171141.1 - 2 414
BC020067.1 - 2 401 3991510

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0