This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNPC3 SIRNA Assays ›
Gene Symbol:
Rnpc3
Gene Name:
RNA-binding region (RNP1, RRM) containing 3
Gene Aliases:
2810441O16Rik, AI447568, C030014B17Rik
Chromosome Location:
Chr.3: 113605067 - 113630149 on Build GRCm38
UniGene ID:
Mm.316928
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 79599

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnpc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038696.1 NP_001033785.1 10 1178
NM_001287015.1 NP_001273944.1 10 1178
NM_026043.3 NP_080319.2 10 1178
GenBank mRNA AK134241.1 - 10 1143
AK161063.1 - 10 1161
AK161974.1 - 10 1131
AK163729.1 - 10 1131
AK173272.1 - 10 929
BC016603.1 - 7 694 3587176
BC043695.1 - 10 1147 5250774

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
molecular_function
nucleic acid binding
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header