This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CISD2 SIRNA Assays ›
Gene Symbol:
Cisd2
Gene Name:
CDGSH iron sulfur domain 2
Gene Aliases:
1500009M05Rik, 1500026J14Rik, 1500031D15Rik, AI848398, B630006A20Rik, Miner1, Noxp70, Zcd2
Chromosome Location:
Chr.3: 135406412 - 135424442 on Build GRCm38
UniGene ID:
Mm.269993
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 79485

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cisd2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025902.3 NP_080178.1 3 1918
GenBank mRNA AK005184.1 - 3 1879
AM162549.1 - 3 2425
BC025614.1 - 1 998
BC037182.1 - 1 186
BC052543.1 - 1 491
BC058279.1 - 3 1917 6528467

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy
autophagy
multicellular organism aging
regulation of autophagy
protein homodimerization activity
poly(A) RNA binding
metal ion binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0