This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IFT27 SIRNA Assays ›
Gene Symbol:
Ift27
Gene Name:
intraflagellar transport 27
Gene Aliases:
2600013G09Rik, Rabl4
Chromosome Location:
Chr.15: 78159463 - 78174108 on Build GRCm38
UniGene ID:
Mm.30191
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 79415

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ift27
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025931.2 NP_080207.1 5 632
GenBank mRNA AK011196.1 - 5 630
AK044278.1 - 2 1070
BC017514.1 - 5 632 4502942

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
smoothened signaling pathway
small GTPase mediated signal transduction
protein transport
Rab protein signal transduction
intraciliary transport
nucleotide binding
GTPase activity
protein binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0