This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SKA2 SIRNA Assays ›
Gene Symbol:
Ska2
Gene Name:
spindle and kinetochore associated complex subunit 2
Gene Aliases:
1110001A07Rik, C78640, Fam33a
Chromosome Location:
Chr.11: 87109261 - 87122974 on Build GRCm38
UniGene ID:
Mm.45008
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 78055

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ska2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025377.3 NP_079653.1 3 214
GenBank mRNA AK003196.1 - 3 242
AK013317.1 - 3 215
AK013370.1 - 3 215
AK017811.1 - 3 215
AK153219.1 - 185
AK162628.1 - 211
BC052151.1 - 215
BC060088.1 - 217
BC096627.1 - 226
BC117001.1 - 3 244 40126320
BC119167.1 - 3 244 8733916

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic anaphase
cell cycle
chromosome segregation
mitotic nuclear division
regulation of microtubule polymerization or depolymerization
cell division
microtubule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0