This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IER3IP1 SIRNA Assays ›
Gene Symbol:
Ier3ip1
Gene Name:
immediate early response 3 interacting protein 1
Gene Aliases:
1110057H19Rik, AI644142, AL022842, AL022863, AV026606
Chromosome Location:
Chr.18: 76930027 - 76941614 on Build GRCm38
UniGene ID:
Mm.28593
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 77993

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ier3ip1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025409.3 NP_079685.1 2 218
GenBank mRNA AK004281.1 - 2 203
AK007388.1 - 2 166
AK007862.1 - 2 150
AK009116.1 - 2 151
AK182132.1 - 2 88
AK210106.1 - 2 88
BC021512.1 - 2 208 5351700
CX207811.1 - 2 207

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of fibroblast apoptotic process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0