This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 0610009O20RIK SIRNA Assays ›
Gene Symbol:
0610009O20Rik
Gene Name:
RIKEN cDNA 0610009O20 gene
Gene Aliases:
2700004E22Rik, Dele, Kiaa0141
Chromosome Location:
Chr.18: 38250249 - 38262629 on Build GRCm38
UniGene ID:
Mm.281086
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 77386

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 0610009O20Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024179.5 NP_077141.2 3 279
GenBank mRNA AK002437.1 - 3 269
AK035876.1 - 3 274
AK079423.1 - 3 262
AK141675.1 - 241
AK171837.1 - 3 270
AK172900.1 - 3 204
BC002117.1 - 3 253 3490474
BY193394.1 - 3 280

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

extrinsic apoptotic signaling pathway via death domain receptors
regulation of cysteine-type endopeptidase activity involved in apoptotic process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0