This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RSRP1 SIRNA Assays ›
Gene Symbol:
Rsrp1
Gene Name:
arginine/serine rich protein 1
Gene Aliases:
2700043I21Rik, D4Ucla2, D4Wsu53e
Chromosome Location:
Chr.4: 134923625 - 134927370 on Build GRCm38
UniGene ID:
Mm.331964
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 77301

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rsrp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023665.3 NP_076154.3 5 1044
GenBank mRNA AB041555.1 - 6 1775
AK003799.1 - 4 2347
AK028145.1 - 4 2349
AK029529.1 - 4 2345
AK150024.1 - 5 1045
AK150665.1 - 5 1024
AK152736.1 - 5 1024
AK160993.1 - 4 2346
AK169760.1 - 4 2347
AK178549.1 - 4 390
AK187384.1 - 4 392
BC043057.1 - 4 2331 6419597
BC046231.1 - 4 2249 6813388
BC054728.1 - 4 2319 6419723
BC056986.1 - 4 2280 6816649

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014193, BC085888, FQ230600.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0