This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MYL12B SIRNA Assays ›
Gene Symbol:
Myl12b
Gene Name:
myosin, light chain 12B, regulatory
Gene Aliases:
1500001M02Rik, C77744, Mylc2b, RLC-B
Chromosome Location:
Chr.17: 70973963 - 70990516 on Build GRCm38
UniGene ID:
Mm.261329
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 76858

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Myl12b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023402.2 NP_075891.1 4 762
GenBank mRNA AK002885.1 - 4 764
AK005133.1 - 4 804
AK146115.1 - 4 780
AK160123.1 - 4 805
AK168316.1 - 4 763
BC028878.1 - 4 744 4218748
BC099425.1 - 3 733 30847912
BC131925.1 - 4 738 40130202
BC131927.1 - 4 738 40130204

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell shape
platelet aggregation
calcium ion binding
myosin heavy chain binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0