This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LPAR3 SIRNA Assays ›
Gene Symbol:
Lpar3
Gene Name:
lysophosphatidic acid receptor 3
Gene Aliases:
Edg7, lpA3
Chromosome Location:
Chr.3: 146220961 - 146286214 on Build GRCm38
UniGene ID:
Mm.155520
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 76721

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lpar3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022983.4 NP_075359.1 3 1416
GenBank mRNA AF293845.2 - 3 1381
AK042084.1 - 3 1393
AK042255.1 - 3 1405
AK132289.1 - 3 1359

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
signal transduction
G-protein coupled receptor signaling pathway
bleb assembly
positive regulation of MAPK cascade
positive regulation of collateral sprouting
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of calcium ion transport
G-protein alpha-subunit binding
signal transducer activity
G-protein coupled receptor activity
phospholipid binding
lysophosphatidic acid receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0