This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC28A3 SIRNA Assays ›
Gene Symbol:
Slc28a3
Gene Name:
solute carrier family 28 (sodium-coupled nucleoside transporter), member 3
Gene Aliases:
Cnt3
Chromosome Location:
Chr.13: 58553307 - 58610877 on Build GRCm38
UniGene ID:
Mm.18188
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 76470

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc28a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022317.3 NP_071712.3 4 377
GenBank mRNA AF305211.1 - 4 310
AK052784.1 - 4 378
AK145077.1 - 4 378
AK164752.1 - 4 378
BC010472.1 - 4 365 3586005
BC013783.1 - 4 368 3600786
BY736278.1 - 4 378

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
transport
purine nucleobase transport
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
sodium ion transmembrane transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
nucleoside binding
nucleoside transmembrane transporter activity
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0