This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCGB1B2 SIRNA Assays ›
Gene Symbol:
Scgb1b2
Gene Name:
secretoglobin, family 1B, member 2
Gene Aliases:
Abpa2, Abph, Apbh, LGP, Mja1l, Scgb1b1
Chromosome Location:
Chr.7: 31290519 - 31291816 on Build GRCm38
UniGene ID:
Mm.6205
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 75248

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Scgb1b2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020563.3 NP_065588.1 2 246
GenBank mRNA AF008595.1 - 2 247
BC115490.1 - 2 247 40100845
BC115491.1 - 2 247 40100846
GU269233.1 - 2 230

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0