This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HERPUD2 SIRNA Assays ›
Gene Symbol:
Herpud2
Gene Name:
HERPUD family member 2
Gene Aliases:
5031400M07Rik, AB041580
Chromosome Location:
Chr.9: 25108124 - 25151781 on Build GRCm38
UniGene ID:
Mm.142843
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 75176

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Herpud2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020586.2 NP_065611.1 2 979
GenBank mRNA AB041580.1 - 2 774
AK019862.1 - 2 924
AK135935.1 - 2 980
BC029691.1 - 2 197 5375603
BC043693.1 - 2 1018 5150485

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to unfolded protein
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm