This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DCPP2 SIRNA Assays ›
Gene Symbol:
Dcpp2 Dcpp1
Gene Name:
demilune cell and parotid protein 2
demilune cell and parotid protein 1
Gene Aliases:
Dcpp-2, EG630537, SPT-2
BC005655, C86550, Dcpp, Dcpp-1, p20
Chromosome Location:
Chr.17: 23898722 - 23900787 on Build GRCm38
Chromosome Location:
Chr.17: 23880876 - 23882853 on Build GRCm38
UniGene ID:
Mm.436677
Mm.371556
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 74601

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dcpp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC059096.1 - 2 223 4914089
BC171996.1 - 205 9054282
DQ237937.1 - 204
Gene Symbol: Dcpp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC024627.1 - 186 4925312
BC045522.1 - 3 196 4190591
BC090399.1 - 186 4168483
DQ237936.1 - 204
S76879.1 - 245
U03711.1 - 209

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0