This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THSD1 SIRNA Assays ›
Gene Symbol:
Thsd1
Gene Name:
thrombospondin, type I, domain 1
Gene Aliases:
4833423O18Rik, Tmtsp
Chromosome Location:
Chr.8: 22225858 - 22261334 on Build GRCm38
UniGene ID:
Mm.32067
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 74016

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Thsd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205253.1 NP_001192182.1 2 1067
NM_001293688.1 NP_001280617.1 3 1210
NM_019576.2 NP_062522.1 3 1210
GenBank mRNA AB039946.1 - 2 1038
AK049636.1 - 3 1173
AK052555.1 - 2 1008
BC051957.1 - 3 1167
BC119603.2 - 2 1038 40060210

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header