This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UCHL5 SIRNA Assays ›
Gene Symbol:
Uchl5
Gene Name:
ubiquitin carboxyl-terminal esterase L5
Gene Aliases:
5830413B11Rik, Uch37
Chromosome Location:
Chr.1: 143777221 - 143807466 on Build GRCm38
UniGene ID:
Mm.261004
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 74006

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Uchl5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159866.1 NP_001153338.1 9 950
NM_019562.2 NP_062508.2 9 953
GenBank mRNA AF148447.1 - 9 896
AF175903.1 - 9 902
AK007860.1 - 8 764
AK011117.1 - 9 920
AK017925.1 - 9 948
BC006891.1 - 9 903 2650799

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
transcription, DNA-templated
regulation of transcription, DNA-templated
proteolysis
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
negative regulation of endopeptidase activity
protein deubiquitination
lateral ventricle development
midbrain development
forebrain morphogenesis
regulation of proteasomal protein catabolic process
thiol-dependent ubiquitin-specific protease activity
endopeptidase inhibitor activity
peptidase activity
cysteine-type peptidase activity
omega peptidase activity
hydrolase activity
poly(A) RNA binding
proteasome binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0