This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FNBP4 SIRNA Assays ›
Gene Symbol:
Fnbp4
Gene Name:
formin binding protein 4
Gene Aliases:
FBP30, Fnbp30, mKIAA1014
Chromosome Location:
Chr.2: 90745370 - 90781021 on Build GRCm38
UniGene ID:
Mm.489638
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 73467

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fnbp4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018828.2 NP_061298.1 5 930
GenBank mRNA AK012167.1 - 5 743
AK014018.1 - 5 747
AK084018.1 - 5 722
AK129263.1 - 5 827
AK164543.1 - 5 701
U40750.2 - 5 930

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013159, XM_006234505.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header