This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDE1A SIRNA Assays ›
Gene Symbol:
Pde1a
Gene Name:
phosphodiesterase 1A, calmodulin-dependent
Gene Aliases:
AI987702, AW125737
Chromosome Location:
Chr.2: 79834453 - 80196518 on Build GRCm38
UniGene ID:
Mm.40678
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 72596

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009978.1 NP_001009978.1 3, 4 313
NM_001009979.1 NP_001009979.1 3, 4 313
NM_001159582.1 NP_001153054.1 5, 6 815
NM_016744.4 NP_058024.2 4, 5 610
GenBank mRNA AF023529.1 - 4, 5 530
AF159298.1 - 3, 4 313
AK014810.1 - 3, 4 312
AK043647.1 - 4, 5 611
AK050004.1 - 5, 6 676
AY845863.1 - 4, 5 362
AY845864.1 - 3, 4 313
BC090628.1 - 4, 5 578 6842675
U56649.1 - 5, 6 546

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_030871, AF327838.

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
regulation of smooth muscle cell apoptotic process
cGMP catabolic process
regulation of smooth muscle cell proliferation
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
calmodulin binding
phosphoric diester hydrolase activity
hydrolase activity
cGMP binding
metal ion binding
calcium- and calmodulin-regulated 3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0