This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GALNS SIRNA Assays ›
Gene Symbol:
Galns
Gene Name:
galactosamine (N-acetyl)-6-sulfate sulfatase
Gene Aliases:
mFLJ00319
Chromosome Location:
Chr.8: 122578237 - 122611503 on Build GRCm38
UniGene ID:
Mm.1786
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 72587

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Galns
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016722.4 NP_057931.3 10, 11 1290
GenBank mRNA AA793622.1 - 2, 3 210
AF111346.1 - 10, 11 1211
AK159592.1 - 10, 11 1267
AK220245.1 - 10, 11 1192
BC004002.1 - 10, 11 1144 3492664

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001047851.

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
catalytic activity
sulfuric ester hydrolase activity
hydrolase activity
N-acetylgalactosamine-6-sulfatase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header