This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HNRNPDL SIRNA Assays ›
Gene Symbol:
Hnrnpdl
Gene Name:
heterogeneous nuclear ribonucleoprotein D-like
Gene Aliases:
AA407431, AA959857, D5Ertd650e, D5Wsu145e, Hnrnpdl1, Hnrpdl, JKTBP, hnRNP DL, hnRNP-DL
Chromosome Location:
Chr.5: 100033577 - 100039950 on Build GRCm38
UniGene ID:
Mm.389579
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 72556

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hnrnpdl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016690.4 NP_057899.2 4 891
GenBank mRNA AB017020.1 - 4 597
AK011767.1 - 4 580
AK172629.1 - 4 550
BC021374.1 - 4 549 5039077
CX219512.1 - 3 266

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001033696, XM_006250663, XM_006250662, XM_008770024, FQ222460, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of gene expression
mRNA metabolic process
nucleotide binding
nucleic acid binding
DNA binding
double-stranded DNA binding
single-stranded DNA binding
RNA binding
poly(A) binding
poly(G) binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0