This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GALNS SIRNA Assays ›
Gene Symbol:
Galns
Gene Name:
galactosamine (N-acetyl)-6-sulfate sulfatase
Gene Aliases:
mFLJ00319
Chromosome Location:
Chr.8: 122578237 - 122611503 on Build GRCm38
UniGene ID:
Mm.1786
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 72395

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Galns
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193645.1 NP_001180574.1 2 344
NM_016722.4 NP_057931.3 2 344
GenBank mRNA AF111346.1 - 2 265
AK033946.1 - 2 262
AK159592.1 - 2 321
AK220245.1 - 2 246
BC004002.1 - 2 198 3492664
BQ550288.1 - 2 110
BY348800.1 - 2 345

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001047851.

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
catalytic activity
sulfuric ester hydrolase activity
hydrolase activity
N-acetylgalactosamine-6-sulfatase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0