This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALDH1A7 SIRNA Assays ›
Gene Symbol:
Aldh1a7
Gene Name:
aldehyde dehydrogenase family 1, subfamily A7
Gene Aliases:
AI987940, Ahd2-like, Aldh-pb, Aldh1a4, Aldhpb
Chromosome Location:
Chr.19: 20692953 - 20727556 on Build GRCm38
UniGene ID:
Mm.14609
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 70628

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aldh1a7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011921.2 NP_036051.1 13 1773
GenBank mRNA BC032880.1 - 13 1750
BC046315.1 - 13 1747 6314273
BC139412.1 - 13 1750 8862434
BC139413.1 - 13 1750 8862435
U96401.1 - 13 1773

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
retinoic acid metabolic process
oxidation-reduction process
retinal dehydrogenase activity
3-chloroallyl aldehyde dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
benzaldehyde dehydrogenase (NAD+) activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm