This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STK11 SIRNA Assays ›
Gene Symbol:
Stk11
Gene Name:
serine/threonine kinase 11
Gene Aliases:
AA408040, Lkb1, Par-4, R75140
Chromosome Location:
Chr.10: 80115693 - 80130685 on Build GRCm38
UniGene ID:
Mm.44231
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 69552

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stk11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB015801.1 - 8 1106
AF129870.1 - 8 1484
AF145287.1 - 8 1088
AF151711.1 - 8 1088
AK004499.1 - 6 767
AK147802.1 - 8 1148
AK171909.1 - 9 1882
AK172385.1 - 8 1871
AK172528.1 - 8 1868
BC052379.1 - 8 1149 5688695
CD565465.1 - 1 14
EU730638.1 - 8 1067

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0