This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC12A7 SIRNA Assays ›
Gene Symbol:
Slc12a7
Gene Name:
solute carrier family 12, member 7
Gene Aliases:
AA408796, D13Ertd261e, Kcc4
Chromosome Location:
Chr.13: 73733053 - 73816755 on Build GRCm38
UniGene ID:
Mm.275800
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 69400

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc12a7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011390.2 NP_035520.1 2 249
GenBank mRNA AF087436.1 - 2 251
AK052910.1 - 2 300
AK083598.1 - 2 207
AK131129.1 - 2 203
AK143535.1 - 2 243
AK149750.1 - 2 244
AK166215.1 - 2 243
AK169950.1 - 2 239
AK171498.1 - 2 286
BC051558.1 - 2 238
BC053470.1 - 2 875
BC059242.1 - 2 243
BC060133.1 - 2 243
BC079677.1 - 2 207
BC141107.1 - 2 209 9055719

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013144, XM_006227789, BC086339, AY429042, AY387485.

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
cell volume homeostasis
transmembrane transport
chloride transmembrane transport
symporter activity
cation:chloride symporter activity
potassium:chloride symporter activity
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0