This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PON1 SIRNA Assays ›
Gene Symbol:
Pon1
Gene Name:
paraoxonase 1
Gene Aliases:
Pon
Chromosome Location:
Chr.6: 5168090 - 5193987 on Build GRCm38
UniGene ID:
Mm.237657
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 68839

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pon1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011134.3 NP_035264.2 6 847
GenBank mRNA BC012706.1 - 6 720 4158951
U32684.1 - 6 729
U72636.1 - 6 729

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

blood circulation
cholesterol metabolic process
response to toxic substance
positive regulation of cholesterol efflux
dephosphorylation
aromatic compound catabolic process
positive regulation of transporter activity
carboxylic acid catabolic process
organophosphate catabolic process
phosphatidylcholine metabolic process
positive regulation of binding
response to fluoride
aryldialkylphosphatase activity
arylesterase activity
calcium ion binding
phospholipid binding
high-density lipoprotein particle binding
hydrolase activity
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header