This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other P4HA2 SIRNA Assays ›
Gene Symbol:
P4ha2
Gene Name:
procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide
Gene Aliases:
AA407196, C76437, P4hl
Chromosome Location:
Chr.11: 54100924 - 54131668 on Build GRCm38
UniGene ID:
Mm.3705
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 68485

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: P4ha2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136076.2 NP_001129548.1 3 337
NM_011031.2 NP_035161.2 3 337
GenBank mRNA AK053504.1 - 3 298
AK142624.1 - 3 286
AK144124.1 - 3 283
AK145535.1 - 3 264
BC018411.1 - 3 249 3157481
U16163.1 - 2 224

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

peptidyl-proline hydroxylation to 4-hydroxy-L-proline
oxidation-reduction process
procollagen-proline 4-dioxygenase activity
iron ion binding
protein binding
oxidoreductase activity
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
L-ascorbic acid binding
metal ion binding
dioxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0