This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STYX SIRNA Assays ›
Gene Symbol:
STYX
Gene Name:
serine/threonine/tyrosine interacting protein
Gene Aliases:
-
Chromosome Location:
Chr.14: 52730159 - 52774989 on Build GRCh38
UniGene ID:
Hs.364980
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 6798

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: STYX
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130701.1 NP_001124173.1 7 379
NM_145251.3 NP_660294.1 6 627
XM_006720242.3 XP_006720305.1 4 247
XM_011537108.2 XP_011535410.1 6 294
XM_011537109.2 XP_011535411.1 3 188
GenBank mRNA AK098195.1 - 6 610
AL832591.1 - 7 364
BC020265.2 - 6 578 3922829
BC146995.1 - 6 517 9056923
BC146998.1 - 6 517 9056926
BX647636.1 - 6 585

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
spermatogenesis
MAPK export from nucleus
regulation of ERK1 and ERK2 cascade
protein binding
protein tyrosine/serine/threonine phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0