This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ITSN1 SIRNA Assays ›
Gene Symbol:
Itsn1
Gene Name:
intersectin 1 (SH3 domain protein 1A)
Gene Aliases:
AA517634, AA545208, AI316805, AI839402, AI848451, Ehsh1, Ese1, Itsn, Sh3p17
Chromosome Location:
Chr.16: 91729354 - 91920591 on Build GRCm38
UniGene ID:
Mm.40546
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 67630

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Itsn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110275.1 NP_001103745.1 7 869
NM_001110276.1 NP_001103746.1 7 869
NM_010587.2 NP_034717.2 7 869
GenBank mRNA AF132478.1 - 6 608
AF132481.1 - 6 608
AK155397.1 - 7 824
AK160949.1 - 7 870
BC062938.1 - 7 869
BC066105.1 - 7 731

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

endocytosis
small GTPase mediated signal transduction
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of phosphorylation
positive regulation of catalytic activity
negative regulation of neuron apoptotic process
positive regulation of protein kinase B signaling
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
calcium ion binding
protein binding
kinase activator activity
protein complex scaffold
metal ion binding
proline-rich region binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0