This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BCAP29 SIRNA Assays ›
Gene Symbol:
BCAP29
Gene Name:
B-cell receptor-associated protein 29
Gene Aliases:
BAP29
Chromosome Location:
Chr.7: 107579977 - 107623317 on Build GRCh38
UniGene ID:
Hs.303787
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 6628

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BCAP29
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008405.2 NP_001008405.1 4 425
NM_018844.3 NP_061332.2 5 750
NR_027830.1 4 644
XM_006716051.2 XP_006716114.1 5 746
XM_011516406.2 XP_011514708.1 5 509
XM_011516407.1 XP_011514709.1 5 746
XM_011516408.2 XP_011514710.1 5 746
XM_011516409.2 XP_011514711.1 5 746
XM_017012436.1 XP_016867925.1 5 746
XM_017012437.1 XP_016867926.1 5 746
XM_017012438.1 XP_016867927.1 5 746
GenBank mRNA AA826200.1 - 1 46
AF126020.1 - 5 460
AK289401.1 - 5 507
AK294824.1 - 4 375
AK298594.1 - 2 151
AW978976.1 - 1 70
BC008478.1 - 5 462 4294273
BM023571.1 - 2 220
BP261060.1 - 2 89
BQ230678.1 - 27
BT006981.1 - 4 411
BX647653.1 - 5 460
CK820004.1 - 3 282

Back To Top

More Information


Gene Ontology Categories:

Function(s)

osteoblast differentiation
intracellular protein transport
ER to Golgi vesicle-mediated transport
apoptotic process
protein localization to endoplasmic reticulum exit site

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm