This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ENTPD1 SIRNA Assays ›
Gene Symbol:
Entpd1
Gene Name:
ectonucleoside triphosphate diphosphohydrolase 1
Gene Aliases:
2610206B08Rik, AA408691, Cd39, NTPDase-1
Chromosome Location:
Chr.19: 40612365 - 40741602 on Build GRCm38
UniGene ID:
Mm.2824
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 66108

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Entpd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF037366.1 - 3, 4 336
AK029512.1 - 3, 4 382
AK030601.1 - 3, 4 362
BC011278.1 - 3, 4 347 3674983
CN696192.1 - 3, 4 385

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001164181, NM_001098175, NM_001164178, NM_001164179, NM_001776, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0