This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAPNS1 SIRNA Assays ›
Gene Symbol:
Capns1
Gene Name:
calpain, small subunit 1
Gene Aliases:
Capa-4, Capa4, Capn4, D7Ertd146e
Chromosome Location:
Chr.7: 30186942 - 30198794 on Build GRCm38
UniGene ID:
Mm.439782
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 66064

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Capns1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009795.3 NP_033925.2 4 371
GenBank mRNA AF058298.1 - 3 350
AK028092.1 - 2 117
AK132819.1 - 3 181
BC018352.1 - 4 365 3156021
BC090988.1 - 4 371 30246016

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017118, XM_008759263, XM_006228796, XM_003748836, FQ229527, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
calcium-dependent cysteine-type endopeptidase activity
calcium ion binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0