siRNA Details
Gene Symbol: FAM215A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026770.1 | 1 | 572 | |||
GenBank mRNA | AF143236.1 | - | 1 | 585 | ||
BC069097.1 | - | 1 | 572 |

Gene Symbol: FAM215A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026770.1 | 1 | 572 | |||
GenBank mRNA | AF143236.1 | - | 1 | 585 | ||
BC069097.1 | - | 1 | 572 |