This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VMN2R47 SIRNA Assays ›
Gene Symbol:
Vmn2r47 Vmn2r30
Gene Name:
vomeronasal 2, receptor 47
vomeronasal 2, receptor 30
Gene Aliases:
-
ENSMUSG00000070847, V2r15
Chromosome Location:
Chr.7: 9816782 - 9841325 on Build GRCm38
Chromosome Location:
Chr.7: 7312274 - 7337493 on Build GRCm38
UniGene ID:
Mm.465205
Mm.458977
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 65284

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vmn2r47
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105151.1 NP_001098621.1 3 1268
Gene Symbol: Vmn2r30
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009490.3 NP_033516.3 3 1268
GenBank mRNA AF011425.1 - 1307

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0