This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTC3 SIRNA Assays ›
Gene Symbol:
Ttc3
Gene Name:
tetratricopeptide repeat domain 3
Gene Aliases:
2610202A04Rik, AA409221, D16Ium21, D16Ium21e, TPRD, mKIAA4119
Chromosome Location:
Chr.16: 94370739 - 94469222 on Build GRCm38
UniGene ID:
Mm.213408
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 65242

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009441.2 NP_033467.2 2, 3 281
GenBank mRNA AB008516.1 - 1, 2 158
AK053765.1 - 2, 3 385
AK088273.1 - 2, 3 282
AK089974.1 - 2, 3 224
AK166298.1 - 2, 3 223
AK220495.1 - 2, 3 323
BC019173.1 - 2, 3 250
BC057207.1 - 2, 3 239
BC145489.1 - 1, 2 198

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108315, XM_008768582.

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
negative regulation of cell morphogenesis involved in differentiation
negative regulation of neuron differentiation
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0