This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TSPYL1 SIRNA Assays ›
Gene Symbol:
Tspyl1
Gene Name:
testis-specific protein, Y-encoded-like 1
Gene Aliases:
Tspyl
Chromosome Location:
Chr.10: 34282190 - 34284885 on Build GRCm38
UniGene ID:
Mm.20445
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 65140

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tspyl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009433.3 NP_033459.1 1 728
GenBank mRNA AF042180.1 - 1 692
AK053812.1 - 1 679
AK159432.1 - 1 709
AK166438.1 - 1 709
AK166805.1 - 1 657
AK166893.1 - 1 657
BC011213.1 - 1 675 4196841

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_003309, BC048969, CR533535, AL136629, BC068554.

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0