This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THRB SIRNA Assays ›
Gene Symbol:
Thrb
Gene Name:
thyroid hormone receptor beta
Gene Aliases:
Nr1a2, T3R[b], T3Rbeta, Thrb1, Thrb2, c-erbAbeta
Chromosome Location:
Chr.14: 17659218 - 18040097 on Build GRCm38
UniGene ID:
Mm.32563
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 65092

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Thrb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113417.1 NP_001106888.1 6 642
NM_009380.3 NP_033406.1 3 728
GenBank mRNA AB221613.1 - 3 495
AK133662.1 - 3 225
AK158826.1 - 6 643
BC089035.1 - 3 728 30695885
BC119552.1 - 3 665 40057420
BC119553.1 - 3 669 40057421
S62756.1 - 5 512

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
sensory perception of sound
negative regulation of female receptivity
regulation of heart contraction
female courtship behavior
positive regulation of cell proliferation
organ morphogenesis
regulation of lipid metabolic process
intracellular receptor signaling pathway
positive regulation of chondrocyte differentiation
steroid hormone mediated signaling pathway
positive regulation of ossification
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
Type I pneumocyte differentiation
regulation of cholesterol metabolic process
regulation of triglyceride metabolic process
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
enzyme binding
chromatin DNA binding
protein homodimerization activity
sequence-specific DNA binding
metal ion binding
thyroid hormone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0