This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLK SIRNA Assays ›
Gene Symbol:
Slk
Gene Name:
STE20-like kinase
Gene Aliases:
9A2, AV021402, AW411554, Etk4, SMAK, Stk2, mKIAA0204, mSLK
Chromosome Location:
Chr.19: 47580019 - 47645246 on Build GRCm38
UniGene ID:
Mm.281011
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 64649

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164639.1 NP_001158111.1 1, 2 333
NM_009289.3 NP_033315.2 1, 2 333
GenBank mRNA AF039574.1 - 1, 2 338
AF112855.1 - 1, 2 222
AK029491.1 - 1, 2 200
AK122218.1 - 1, 2 674
BC131674.1 - 1, 2 234 40110915
BC131675.1 - 1, 2 234 40110921

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_014720, BC047885, BC047762, AB002804, BC111565, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
regulation of mitotic cell cycle
phosphorylation
signal transduction by protein phosphorylation
stress-activated protein kinase signaling cascade
activation of protein kinase activity
positive regulation of histone phosphorylation
regulation of apoptotic process
positive regulation of apoptotic process
protein autophosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0