This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PROX1 SIRNA Assays ›
Gene Symbol:
Prox1
Gene Name:
prospero homeobox 1
Gene Aliases:
A230003G05Rik
Chromosome Location:
Chr.1: 190118035 - 190170825 on Build GRCm38
UniGene ID:
Mm.132579
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 63890

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prox1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008937.2 NP_032963.1 2 601
GenBank mRNA AF061576.1 - 1 188
AK052889.1 - 2 635
AK143179.1 - 2 425
BC051411.1 - 2 601 6490801

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107201, XM_006250456, XM_006250454, XM_006250455, XM_006250458.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
cell fate determination
positive regulation of endothelial cell proliferation
lymph vessel development
lymphangiogenesis
lens morphogenesis in camera-type eye
hepatocyte cell migration
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
positive regulation of cell proliferation
negative regulation of cell proliferation
regulation of gene expression
positive regulation of endothelial cell migration
dentate gyrus development
cerebellar granule cell differentiation
skeletal muscle thin filament assembly
regulation of circadian rhythm
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of viral genome replication
endothelial cell differentiation
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
inner ear development
venous blood vessel morphogenesis
ventricular cardiac myofibril assembly
atrial cardiac muscle tissue morphogenesis
ventricular cardiac muscle tissue morphogenesis
endocardium formation
positive regulation of sarcomere organization
ventricular septum morphogenesis
aorta smooth muscle tissue morphogenesis
positive regulation of heart growth
lymphatic endothelial cell differentiation
regulation of transcription involved in lymphatic endothelial cell fate commitment
branching involved in pancreas morphogenesis
lens fiber cell morphogenesis
negative regulation of bile acid biosynthetic process
hepatocyte proliferation
acinar cell differentiation
neuronal stem cell population maintenance
positive regulation of cell cycle checkpoint
positive regulation of neural precursor cell proliferation
positive regulation of forebrain neuron differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein binding
ligand-dependent nuclear receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
DBD domain binding
LBD domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0