This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARHGEF1 SIRNA Assays ›
Gene Symbol:
Arhgef1
Gene Name:
Rho guanine nucleotide exchange factor (GEF) 1
Gene Aliases:
Lbcl2, Lsc
Chromosome Location:
Chr.7: 24902917 - 24926594 on Build GRCm38
UniGene ID:
Mm.3181
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 62783

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arhgef1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130150.1 NP_001123622.1 3, 4 271
NM_001130151.1 NP_001123623.1 3, 4 271
NM_001130152.1 NP_001123624.1 3, 4 280
NM_001130153.1 NP_001123625.1 3, 4 271
NM_008488.2 NP_032514.1 3, 4 271
GenBank mRNA AK012156.1 - 3, 4 221
AK043027.1 - 3, 4 235
AK051518.1 - 3, 4 221
AK131181.1 - 3, 4 168
AK157056.1 - 3, 4 207
AK162482.1 - 3, 4 184
AK172354.1 - 3, 4 207
AK211978.1 - 1, 2 39
BB840025.1 - 3, 4 271
BC012488.1 - 3, 4 231 3154558
BY323306.1 - 3, 4 281
CV783191.1 - 3, 4 203
U58203.1 - 3, 4 201

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: AF314539.

Gene Ontology Categories:

Function(s) Process(es)

regulation of Rho protein signal transduction
termination of G-protein coupled receptor signaling pathway
negative regulation of cell death
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header