This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IFNGR2 SIRNA Assays ›
Gene Symbol:
Ifngr2
Gene Name:
interferon gamma receptor 2
Gene Aliases:
Ifgr2, Ifgt
Chromosome Location:
Chr.16: 91547094 - 91564007 on Build GRCm38
UniGene ID:
Mm.249364
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 62292

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ifngr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008338.3 NP_032364.1 2, 3 266
GenBank mRNA AK086929.1 - 2, 3 280
AK137165.1 - 2, 3 289
AK143214.1 - 2, 3 281
AK146995.1 - 284
AK165193.1 - 2, 3 282
AK169907.1 - 2, 3 286
BC055745.1 - 2, 3 266 6403454
S69336.1 - 2, 3 290

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytokine-mediated signaling pathway
cytokine receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0