This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IFIT1 SIRNA Assays ›
Gene Symbol:
Ifit1
Gene Name:
interferon-induced protein with tetratricopeptide repeats 1
Gene Aliases:
ISG56, Ifi56, P56
Chromosome Location:
Chr.19: 34640889 - 34650009 on Build GRCm38
UniGene ID:
Mm.439751
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 62197

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ifit1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008331.3 NP_032357.2 2 228
GenBank mRNA AK089608.1 - 2 235
AK131669.1 - 2 225
AK144230.1 - 2 233
AK150352.1 - 2 235
AK150424.1 - 2 229
AK150609.1 - 2 238
AK151125.1 - 2 238
AK151158.1 - 2 229
AK151278.1 - 2 238
AK151298.1 - 2 232
AK151983.1 - 2 231
AK152112.1 - 2 231
AK152753.1 - 2 229
AK153184.1 - 2 231
AK153500.1 - 2 229
AK153532.1 - 2 238
AK157848.1 - 2 229
BC003768.1 - 2 212 3496673
CT010226.1 - 1 134
U43084.1 - 2 203

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_020096, AJ276893.

Gene Ontology Categories:

Function(s) Process(es)

immune system process
response to virus
cellular response to interferon-alpha
cellular response to interferon-beta
innate immune response
defense response to virus
RNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0