This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAS SIRNA Assays ›
Gene Symbol:
Fas
Gene Name:
Fas (TNF receptor superfamily member 6)
Gene Aliases:
AI196731, APO1, APT1, CD95, TNFR6, Tnfrsf6, lpr
Chromosome Location:
Chr.19: 34290659 - 34327775 on Build GRCm38
UniGene ID:
Mm.1626
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 61450

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fas
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146708.1 NP_001140180.1 2 218
NM_007987.2 NP_032013.2 2 218
GenBank mRNA AK002590.1 - 2 169
AK086933.1 - 2 195
BC061160.1 - 2 191 30302649
BY348234.1 - 2 219
DQ846748.1 - 2 162
DQ846749.1 - 2 162
L25336.1 - 1 141
M83649.1 - 2 211

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immunoglobulin production
renal system process
apoptotic process
activation-induced cell death of T cells
inflammatory cell apoptotic process
obsolete transformed cell apoptotic process
immune response
signal transduction
extrinsic apoptotic signaling pathway via death domain receptors
response to toxic substance
gene expression
regulation of gene expression
positive regulation of cell death
B cell mediated immunity
dendrite regeneration
positive regulation of protein homooligomerization
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
regulation of apoptotic process
T cell homeostasis
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative thymic T cell selection
regulation of lymphocyte differentiation
regulation of myeloid cell differentiation
spleen development
negative regulation of B cell activation
protein homooligomerization
response to glucocorticoid
neuron apoptotic process
cellular response to lithium ion
cellular response to hyperoxia
positive regulation of release of cytochrome c from mitochondria
motor neuron apoptotic process
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
hepatocyte apoptotic process
necroptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
protease binding
transmembrane signaling receptor activity
tumor necrosis factor-activated receptor activity
protein binding
kinase binding
protein complex binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0