This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EI24 SIRNA Assays ›
Gene Symbol:
Ei24
Gene Name:
etoposide induced 2.4 mRNA
Gene Aliases:
AA536736, AI115355, PIG8
Chromosome Location:
Chr.9: 36779153 - 36797393 on Build GRCm38
UniGene ID:
Mm.4337
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 61115

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ei24
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199494.1 NP_001186423.1 7 729
NM_007915.5 NP_031941.1 7 613
GenBank mRNA AK145018.1 - 7 690
AK147776.1 - 7 642
AK159927.1 - 7 671
AK172233.1 - 7 693
AK212828.1 - 2 174
BU148129.1 - 7 682
U41751.2 - 7 617

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001025660, XM_006242801, XM_006242800, BC098721.

Gene Ontology Categories:

Function(s) Process(es)

autophagy
apoptotic process
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of cell growth
response to drug
neuromuscular process controlling balance
cellular response to UV-C
positive regulation of intrinsic apoptotic signaling pathway
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0