This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DIO1 SIRNA Assays ›
Gene Symbol:
Dio1
Gene Name:
deiodinase, iodothyronine, type I
Gene Aliases:
5DI, D1, ITDI1, TXDI1
Chromosome Location:
Chr.4: 107291465 - 107307143 on Build GRCm38
UniGene ID:
Mm.148342
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 61062

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dio1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007860.3 NP_031886.3 1 136
GenBank mRNA AK002549.1 - 1 137
BC019786.1 - 1 161 5133568
BC037080.1 - 1 172
BC089608.1 - 1 136 6766076
U49861.1 - 1 136

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

hormone biosynthetic process
oxidation-reduction process
thyroxine 5'-deiodinase activity
selenium binding
oxidoreductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm