This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CST6 SIRNA Assays ›
Gene Symbol:
Cst6
Gene Name:
cystatin E/M
Gene Aliases:
-
Chromosome Location:
Chr.1: 220727292 - 220729000 on Build Rnor_6.0
UniGene ID:
Rn.9609
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 59522

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cst6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133566.1 NP_598250.1 1 445
GenBank mRNA AY046415.1 - 1 445

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028623, AK078116, AK003744, BC027680, BC061036.

Gene Ontology Categories:

Function(s) Process(es)

epidermis development
negative regulation of endopeptidase activity
cysteine-type endopeptidase inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0