This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IL11 SIRNA Assays ›
Gene Symbol:
Il11
Gene Name:
interleukin 11
Gene Aliases:
Il-11
Chromosome Location:
Chr.1: 72636959 - 72643255 on Build Rnor_6.0
UniGene ID:
Rn.198483
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 59521

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Il11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133519.4 NP_598203.4 2, 3 299
GenBank mRNA AF347935.1 - 2, 3 316

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008350, BC134354, AK132897, BY322682, U03421.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell proliferation
positive regulation of peptidyl-serine phosphorylation
positive regulation of MAPK cascade
positive regulation of transcription from RNA polymerase II promoter
negative regulation of hormone secretion
positive regulation of peptidyl-tyrosine phosphorylation
regulation of complement-dependent cytotoxicity
negative regulation of endothelial cell apoptotic process
cytokine activity
interleukin-11 receptor binding
growth factor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0