This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYTL5 SIRNA Assays ›
Gene Symbol:
Sytl5
Gene Name:
synaptotagmin-like 5
Gene Aliases:
-
Chromosome Location:
Chr.X: 14017405 - 14109592 on Build Rnor_6.0
UniGene ID:
Rn.92480
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 59046

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sytl5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178333.1 NP_848016.1 2 185
XM_008773069.1 XP_008771291.1 4 1515
XM_008773070.1 XP_008771292.1 4 1515
GenBank mRNA AB098162.1 - 2 185

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
exocytosis
Rab GTPase binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0