This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP1R17 SIRNA Assays ›
Gene Symbol:
Ppp1r17
Gene Name:
protein phosphatase 1, regulatory subunit 17
Gene Aliases:
Gsbs
Chromosome Location:
Chr.4: 86268994 - 86292435 on Build Rnor_6.0
UniGene ID:
Rn.24323
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58671

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppp1r17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153467.1 NP_703197.1 3, 4 380
XM_006236545.2 XP_006236607.1 3, 4 889
XM_008762878.1 XP_008761100.1 4, 5 816
GenBank mRNA AF294688.1 - 3, 4 380
FQ213688.1 - 3, 4 342

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of phosphatase activity
negative regulation of catalytic activity
protein phosphatase inhibitor activity
protein serine/threonine phosphatase inhibitor activity
phosphatase inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0