This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB8B SIRNA Assays ›
Gene Symbol:
Rab8b
Gene Name:
RAB8B, member RAS oncogene family
Gene Aliases:
-
Chromosome Location:
Chr.8: 72641680 - 72714646 on Build Rnor_6.0
UniGene ID:
Rn.10995
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58667

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab8b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153317.1 NP_695229.1 6, 7 554
GenBank mRNA BC158579.1 - 6, 7 580 9091689
U53475.1 - 6, 7 554

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016530, AB038995, AK001111, BC020654, AL833365.
This siRNA also targets the following transcripts in Mouse: NM_173413, AK031251, BC059208, AK155621, AK086085, and others.

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle docking involved in exocytosis
metabolic process
protein secretion
synaptic vesicle exocytosis
regulation of exocytosis
antigen processing and presentation
positive regulation of cell projection organization
Rab protein signal transduction
cellular response to insulin stimulus
adherens junction organization
cilium assembly
protein import into peroxisome membrane
Golgi vesicle fusion to target membrane
positive regulation of corticotropin secretion
protein localization to plasma membrane
GTPase activity
receptor binding
protein binding
GTP binding
GDP binding
TPR domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header